Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_079420.2 | NP_524144.1 | 7 | 822 | ||
NM_079422.2 | NP_524146.1 | 7 | 632 | |||
GenBank mRNA | BC005318.1 | - | 7 | 632 | 3933998 | |
M20643.1 | - | 7 | 609 | |||
X05451.1 | - | 7 | 614 |