Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 17 | 2415 | ||
XM_017024011.1 | XP_016879500.1 | 17 | 2380 | |||
XM_017024012.1 | XP_016879501.1 | 17 | 2484 | |||
XM_017024013.1 | XP_016879502.1 | 17 | 2304 | |||
XM_017024014.1 | XP_016879503.1 | 17 | 2904 | |||
XM_017024015.1 | XP_016879504.1 | 14 | 1927 | |||
GenBank mRNA | AF423307.1 | - | 16 | 2272 | ||
AK057068.1 | - | 17 | 2347 | |||
AK074491.1 | - | 10 | 1224 | |||
AY028899.1 | - | 17 | 2415 | |||
CR627382.1 | - | 16 | 2221 |