Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127441.1 | NP_001120913.1 | 11, 12 | 1466 | ||
NM_001127442.1 | NP_001120914.1 | 9, 10 | 1355 | |||
NM_001318223.1 | NP_001305152.1 | 9, 10 | 1355 | |||
NM_080385.4 | NP_525124.3 | 10, 11 | 1413 | |||
XM_005250710.1 | XP_005250767.1 | 10, 11 | 1394 | |||
XM_005250712.1 | XP_005250769.1 | 9, 10 | 1308 | |||
XM_011516698.1 | XP_011515000.1 | 9, 10 | 2031 | |||
XM_011516701.1 | XP_011515003.1 | 9, 10 | 1948 | |||
XM_011516702.1 | XP_011515004.1 | 9, 10 | 1934 | |||
XM_011516703.1 | XP_011515005.1 | 9, 10 | 1927 | |||
XM_011516704.1 | XP_011515006.1 | 9, 10 | 2045 | |||
GenBank mRNA | AF384667.1 | - | 8, 9 | 846 | ||
AK093288.1 | - | 9, 10 | 1068 | |||
AK131191.1 | - | 9, 10 | 1291 | |||
AK315256.1 | - | 9, 10 | 1329 | |||
AY155228.1 | - | 4, 5 | 368 | |||
AY155229.1 | - | 10, 11 | 1366 | |||
AY155230.1 | - | 11, 12 | 1436 | |||
BC039362.1 | - | 11, 12 | 1440 | 5270617 | ||
BC042996.1 | - | 9, 10 | 1982 | 5296768 |