Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012296.3 | NP_036428.1 | 10 | 3669 | ||
NM_080491.2 | NP_536739.1 | 10 | 3725 | |||
XM_006718753.2 | XP_006718816.1 | 11 | 3770 | |||
XM_011545408.2 | XP_011543710.1 | 9 | 3376 | |||
GenBank mRNA | AB011143.1 | - | 10 | 3669 | ||
AB018413.1 | - | 10 | 3623 | |||
BC152459.1 | - | 10 | 3669 | 8862595 |