Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TJAP1 SIRNA Assays ›
Gene Symbol:
TJAP1
Gene Name:
tight junction associated protein 1
Gene Aliases:
PILT, TJP4
Chromosome Location:
Chr.6: 43477523 - 43506557 on Build GRCh38
UniGene ID:
Hs.520145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36114
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TJAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146016.1
NP_001139488.1
11
2374
NM_001146017.1
NP_001139489.1
10
2339
NM_001146018.1
NP_001139490.1
11
2401
NM_001146019.1
NP_001139491.1
10
2344
NM_001146020.1
NP_001139492.1
9
2195
NM_080604.2
NP_542171.2
10
2396
NR_027394.1
10
2261
XM_006715249.1
XP_006715312.1
11
2352
XM_006715250.3
XP_006715313.1
12
2371
XM_006715251.2
XP_006715314.1
11
2379
XM_006715252.2
XP_006715315.1
12
2439
XM_006715254.3
XP_006715317.1
11
2277
XM_006715256.1
XP_006715319.1
11
2274
XM_006715257.2
XP_006715320.1
11
2328
XM_006715259.3
XP_006715322.1
10
2178
XM_006715260.1
XP_006715323.1
11
2295
XM_006715261.1
XP_006715324.1
10
2199
XM_006715262.2
XP_006715325.1
11
2524
XM_006715263.2
XP_006715326.1
10
2234
XM_006715265.1
XP_006715328.1
10
2244
XM_006715266.1
XP_006715329.1
9
2152
XM_006715267.2
XP_006715330.1
11
2363
XM_006715268.2
XP_006715331.1
11
2360
XM_006715269.2
XP_006715332.1
11
2415
XM_006715270.1
XP_006715333.1
10
2333
XM_011514995.1
XP_011513297.1
12
2414
XM_011514996.1
XP_011513298.1
11
2334
XM_017011488.1
XP_016866977.1
10
2252
XM_017011489.1
XP_016866978.1
11
2549
XM_017011490.1
XP_016866979.1
10
2322
XM_017011491.1
XP_016866980.1
11
2358
XM_017011492.1
XP_016866981.1
12
2487
XM_017011493.1
XP_016866982.1
12
2485
GenBank mRNA
AJ420477.1
-
2
1495
AK024269.1
-
10
2346
AK074241.1
-
5
1790
AK094907.1
-
3
2202
AK123398.1
-
7
4904
AK127338.1
-
9
3457
AL117398.1
-
4
2828
BC046239.1
-
11
2294
4906152
BC064401.1
-
10
2275
5539930
Back To Top
More Information
Panther Classification:
Molecular Function -
tight junction
Gene Ontology Categories:
Function(s)
Process(es)
Golgi organization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline