Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 10, 11 | 1998 | ||
NM_080616.4 | NP_542183.2 | 7, 8 | 1337 | |||
XM_005260282.4 | XP_005260339.1 | 10, 11 | 2284 | |||
XM_005260283.4 | XP_005260340.1 | 11, 12 | 2308 | |||
XM_005260284.4 | XP_005260341.1 | 10, 11 | 1583 | |||
XM_005260285.4 | XP_005260342.2 | 10, 11 | 2288 | |||
XM_005260286.4 | XP_005260343.1 | 10, 11 | 2208 | |||
XM_005260288.2 | XP_005260345.1 | 7, 8 | 1385 | |||
XM_005260289.4 | XP_005260346.1 | 7, 8 | 1336 | |||
XM_006723702.3 | XP_006723765.1 | 11, 12 | 1675 | |||
XM_006723703.3 | XP_006723766.1 | 10, 11 | 1587 | |||
XM_017027669.1 | XP_016883158.1 | 12, 13 | 2329 | |||
GenBank mRNA | AK056286.1 | - | 7, 8 | 1140 | ||
BC065370.1 | - | 6, 7 | 1109 | 6261963 |