Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023010.3 | NP_075386.1 | 7 | 897 | ||
NM_080632.2 | NP_542199.1 | 7 | 897 | |||
XM_017029737.1 | XP_016885226.1 | 7 | 897 | |||
XM_017029738.1 | XP_016885227.1 | 7 | 897 | |||
XM_017029739.1 | XP_016885228.1 | 7 | 897 | |||
XM_017029740.1 | XP_016885229.1 | 7 | 897 | |||
GenBank mRNA | AF318576.1 | - | 7 | 817 | ||
AK093728.1 | - | 7 | 835 | |||
AY013251.1 | - | 7 | 867 | |||
BC121017.1 | - | 7 | 840 | 40117102 |