Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080668.3 | NP_542399.1 | 5 | 470 | ||
XM_005273733.3 | XP_005273790.1 | 5 | 465 | |||
XM_005273734.3 | XP_005273791.1 | 5 | 465 | |||
XM_011544743.2 | XP_011543045.1 | 5 | 465 | |||
XM_011544748.2 | XP_011543050.1 | 5 | 465 | |||
XM_011544749.2 | XP_011543051.1 | 5 | 465 | |||
XM_017017158.1 | XP_016872647.1 | 5 | 465 | |||
GenBank mRNA | AK093670.1 | - | 1 | 45 | ||
AK291490.1 | - | 5 | 394 | |||
AL832526.1 | - | 4 | 187 | |||
BC011000.2 | - | 5 | 378 | 3938081 |