Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004395.3 | NP_004386.2 | 3 | 336 | ||
NM_080881.2 | NP_543157.1 | 4 | 452 | |||
XM_005265827.3 | XP_005265884.1 | 3 | 283 | |||
XM_006714826.2 | XP_006714889.1 | 3 | 173 | |||
XM_011534446.2 | XP_011532748.1 | 4 | 455 | |||
XM_011534447.2 | XP_011532749.1 | 3 | 952 | |||
XM_017009139.1 | XP_016864628.1 | 3 | 454 | |||
XM_017009140.1 | XP_016864629.1 | 3 | 445 | |||
GenBank mRNA | AK094125.1 | - | 3 | 267 | ||
AK225523.1 | - | 3 | 277 | |||
AK314645.1 | - | 3 | 277 | |||
AL110225.1 | - | 4 | 781 | |||
BC000283.1 | - | 3 | 260 | 3356428 | ||
BC007281.1 | - | 3 | 260 | 3343626 | ||
BC007567.1 | - | 3 | 260 | 3028196 | ||
BF205663.1 | - | 308 | ||||
D17530.1 | - | 3 | 261 | |||
DA245346.1 | - | 3 | 336 | |||
U00802.1 | - | 3 | 236 |