Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130767.2 | NP_570123.1 | 2, 3 | 249 | ||
XM_006714531.2 | XP_006714594.1 | 2, 3 | 214 | |||
XM_006714532.3 | XP_006714595.1 | 2, 3 | 214 | |||
XM_017009045.1 | XP_016864534.1 | 2, 3 | 214 | |||
XM_017009046.1 | XP_016864535.1 | 2, 3 | 214 | |||
XM_017009047.1 | XP_016864536.1 | 2, 3 | 254 | |||
GenBank mRNA | AB078619.1 | - | 2, 3 | 217 | ||
AK122960.1 | - | 2, 3 | 214 | |||
BC075010.2 | - | 2, 3 | 215 | 30915616 | ||
BC075011.2 | - | 2, 3 | 210 | 30915403 | ||
BC089437.1 | - | 2, 3 | 259 |