Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039350.2 | NP_001034439.1 | 7, 8 | 1154 | ||
NM_001936.4 | NP_001927.3 | 7, 8 | 856 | |||
NM_130797.3 | NP_570629.2 | 7, 8 | 1162 | |||
XM_017011808.1 | XP_016867297.1 | 8, 9 | 1108 | |||
XM_017011809.1 | XP_016867298.1 | 8, 9 | 871 | |||
XM_017011810.1 | XP_016867299.1 | 8, 9 | 974 | |||
XM_017011811.1 | XP_016867300.1 | 8, 9 | 898 | |||
XM_017011812.1 | XP_016867301.1 | 3, 4 | 269 | |||
GenBank mRNA | BC150304.1 | - | 7, 8 | 1153 | 8860401 | |
BX647966.1 | - | 7, 8 | 919 | |||
M96859.1 | - | 7, 8 | 903 | |||
M96860.1 | - | 7, 8 | 750 |