Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270614.1 | NP_001257543.1 | 4, 5 | 699 | ||
NM_001270615.1 | NP_001257544.1 | 4, 5 | 766 | |||
NM_133328.3 | NP_579874.1 | 4, 5 | 781 | |||
NR_073047.1 | 3, 4 | 333 | ||||
NR_073048.1 | 3, 4 | 415 | ||||
NR_073050.1 | 3, 4 | 400 | ||||
GenBank mRNA | AF443591.1 | - | 3, 4 | 582 | ||
AF457575.1 | - | 3, 4 | 582 | |||
AK075328.1 | - | 4, 5 | 705 | |||
AY125488.1 | - | 4, 5 | 650 | |||
BC013372.2 | - | 4, 5 | 642 | 3941907 | ||
BC027930.1 | - | 4, 5 | 650 | 5199772 | ||
BM908148.1 | - | 3, 4 | 327 | |||
BX374729.2 | - | 4, 5 | 666 | |||
BX458265.2 | - | 3, 4 | 366 | |||
CN373297.1 | - | 3, 4 | 372 | |||
HY033487.1 | - | 3, 4 | 324 |