Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122853.2 | NP_001116325.1 | 7 | 1441 | ||
NM_133371.4 | NP_588612.2 | 7 | 1093 | |||
GenBank mRNA | AF052497.1 | - | 2 | 901 | ||
AF480443.1 | - | 7 | 945 | |||
AJ300586.1 | - | 7 | 1056 | |||
AJ300587.1 | - | 7 | 798 | |||
BC069380.1 | - | 7 | 923 | 7262195 | ||
BC069553.1 | - | 7 | 923 | 7262207 |