Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL10L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080746.2 | NP_542784.1 | 1 | 744 | ||
GenBank mRNA | AB063608.1 | - | 1 | 693 | ||
AK130863.1 | - | 1 | 704 | |||
BC014310.1 | - | 1 | 744 | 3935452 | ||
BC066312.1 | - | 1 | 700 | 5296757 |