Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207076.2 | NP_001194005.1 | 4 | 451 | ||
NM_080834.3 | NP_543024.2 | 4 | 451 | |||
NR_130714.1 | 4 | 451 | ||||
XM_005260295.2 | XP_005260352.1 | 4 | 425 | |||
XM_011528590.2 | XP_011526892.1 | 4 | 411 | |||
XM_011528592.1 | XP_011526894.1 | 3 | 281 | |||
XM_017027683.1 | XP_016883172.1 | 4 | 629 | |||
GenBank mRNA | AK057454.1 | - | 4 | 425 | ||
BC114634.1 | - | 4 | 409 | |||
BC114945.1 | - | 4 | 409 | 40084218 | ||
DB023698.1 | - | 4 | 425 | |||
DB056200.1 | - | 5 | 463 | |||
HY023889.1 | - | 4 | 451 | |||
HY055403.1 | - | 4 | 325 |