Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324081.1 | NP_001311010.1 | 6, 7 | 787 | ||
NM_080861.3 | NP_543137.2 | 6, 7 | 809 | |||
XM_005255673.4 | XP_005255730.1 | 6, 7 | 999 | |||
GenBank mRNA | AF403028.1 | - | 5, 6 | 718 | ||
AK057850.1 | - | 567 | ||||
AK074190.1 | - | 6, 7 | 773 | |||
BC007588.1 | - | 6, 7 | 755 | 3342825 | ||
BC024244.1 | - | 6, 7 | 773 | 4798873 | ||
BC035065.1 | - | 6, 7 | 766 | |||
BC041897.1 | - | 5, 6 | 1583 | 5298729 | ||
BC047441.1 | - | 6, 7 | 751 | 5303706 | ||
BC065515.1 | - | 3, 4 | 1052 | |||
BI768095.1 | - | 6, 7 | 763 | |||
BQ930544.1 | - | 1, 2 | 91 |