Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080863.4 | NP_543139.4 | 4, 5 | 1268 | ||
GenBank mRNA | AK054727.1 | - | 4, 5 | 1268 | ||
AK314595.1 | - | 4, 5 | 1260 | |||
AY557346.1 | - | 4, 5 | 894 | |||
BC075088.2 | - | 4, 5 | 1217 | 30915388 | ||
BC118584.1 | - | 4, 5 | 960 | 40084995 | ||
BC118585.1 | - | 4, 5 | 960 | 40084992 |