Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPP8 SIRNA Assays ›
Gene Symbol:
DPP8
Gene Name:
dipeptidyl peptidase 8
Gene Aliases:
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location:
Chr.15: 65442463 - 65517713 on Build GRCh38
UniGene ID:
Hs.458609
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
36294
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPP8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320875.1
NP_001307804.1
3
569
NM_001320876.1
NP_001307805.1
2
538
NM_017743.5
NP_060213.2
2
433
NM_130434.4
NP_569118.1
2
433
NM_197960.3
NP_932064.1
3
474
NM_197961.3
NP_932065.1
3
569
NR_135485.1
2
433
NR_135486.1
3
474
XM_011521730.1
XP_011520032.1
2
1822
XM_011521731.1
XP_011520033.1
2
1822
XM_011521732.2
XP_011520034.1
2
1822
XM_011521733.1
XP_011520035.1
2
414
XM_011521734.1
XP_011520036.1
2
1822
XM_017022373.1
XP_016877862.1
2
1822
XM_017022374.1
XP_016877863.1
2
1822
XM_017022375.1
XP_016877864.1
2
414
XM_017022376.1
XP_016877865.1
2
414
XM_017022377.1
XP_016877866.1
2
414
XM_017022378.1
XP_016877867.1
2
1822
XM_017022379.1
XP_016877868.1
2
414
XM_017022380.1
XP_016877869.1
2
414
XM_017022381.1
XP_016877870.1
2
414
GenBank mRNA
AF221634.1
-
2
407
AK000290.1
-
2
356
AK291057.1
-
3
544
AK296915.1
-
2
509
AK302638.1
-
3
410
AY172659.1
-
1
201
AY354202.1
-
3
402
BC030688.2
-
3
560
4822550
BI458620.1
-
3
432
BI462092.1
-
3
415
DA297112.1
-
2
419
DC401507.1
-
3
475
DC412877.1
-
2
435
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
immune response
serine-type peptidase activity
dipeptidyl-peptidase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0