Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130446.2 | NP_569713.2 | 2 | 384 | ||
XM_011513274.2 | XP_011511576.1 | 2 | 385 | |||
GenBank mRNA | AF441792.1 | - | 2 | 296 | ||
AK097125.1 | - | 2 | 338 | |||
BC032348.1 | - | 2 | 362 | 5213743 | ||
BC037989.1 | - | 2 | 248 | |||
BP284594.1 | - | 2 | 383 | |||
BX648370.1 | - | 2 | 339 |