Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039350.2 | NP_001034439.1 | 8 | 1243 | ||
NM_001290252.1 | NP_001277181.1 | 6 | 810 | |||
NM_001936.4 | NP_001927.3 | 8 | 945 | |||
NM_130797.3 | NP_570629.2 | 8 | 1251 | |||
XM_017011808.1 | XP_016867297.1 | 9 | 1197 | |||
XM_017011809.1 | XP_016867298.1 | 9 | 960 | |||
XM_017011810.1 | XP_016867299.1 | 9 | 1063 | |||
XM_017011811.1 | XP_016867300.1 | 9 | 987 | |||
XM_017011812.1 | XP_016867301.1 | 4 | 358 | |||
GenBank mRNA | AK294483.1 | - | 6 | 580 | ||
BC150304.1 | - | 8 | 1242 | 8860401 | ||
BX647966.1 | - | 8 | 1008 | |||
M96859.1 | - | 8 | 992 | |||
M96860.1 | - | 8 | 839 |