Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130807.2 | NP_570719.1 | 3 | 676 | ||
XM_011527683.2 | XP_011525985.1 | 3 | 753 | |||
GenBank mRNA | AB061793.1 | - | 2 | 539 | ||
AJ580635.1 | - | 1 | 517 | |||
AK095471.1 | - | 3 | 676 | |||
AL832474.1 | - | 3 | 712 | |||
BC015049.1 | - | 2 | 565 | 3919665 |