Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYX1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033559.2 | NP_001028731.1 | 3 | 526 | ||
NM_001033560.1 | NP_001028732.1 | 3 | 526 | |||
NM_130810.3 | NP_570722.2 | 3 | 526 | |||
GenBank mRNA | AF337549.1 | - | 3 | 526 | ||
AK095201.1 | - | 2 | 200 | |||
AK302316.1 | - | 2 | 170 | |||
BC062564.1 | - | 2 | 177 | 5163101 | ||
KU178882.1 | - | 2 | 158 | |||
KU178883.1 | - | 2 | 158 | |||
KU178884.1 | - | 2 | 158 |
Gene Symbol: DYX1C1-CCPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037923.1 | 2 | 413 | |||
GenBank mRNA | AK295788.1 | - | 2 | 170 |