Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032108.3 | NP_115484.2 | 3 | 451 | ||
XM_011527639.2 | XP_011525941.1 | 3 | 500 | |||
XM_011527640.2 | XP_011525942.1 | 3 | 243 | |||
XM_011527641.1 | XP_011525943.1 | 3 | 500 | |||
GenBank mRNA | AB022433.1 | - | 2 | 227 | ||
AB209278.1 | - | 2 | 213 | |||
AF216389.1 | - | 2 | 192 | |||
AF293363.1 | - | 2 | 192 | |||
AK300037.1 | - | 3 | 451 | |||
AY358939.1 | - | 2 | 207 | |||
BC142617.1 | - | 3 | 394 | 40147342 | ||
DA818632.1 | - | 3 | 451 | |||
DB257858.1 | - | 3 | 501 | |||
FR839673.1 | - | 2 | 189 |