Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146225.1 | NP_001139697.1 | 10 | 1825 | ||
NM_001146226.1 | NP_001139698.1 | 8 | 1826 | |||
NM_080747.2 | NP_542785.1 | 9 | 1952 | |||
XM_017018833.1 | XP_016874322.1 | 11 | 1755 | |||
XM_017018834.1 | XP_016874323.1 | 11 | 1693 | |||
XM_017018835.1 | XP_016874324.1 | 11 | 1680 | |||
XM_017018836.1 | XP_016874325.1 | 10 | 1654 | |||
XM_017018837.1 | XP_016874326.1 | 8 | 1288 | |||
GenBank mRNA | AK093060.1 | - | 10 | 1729 | ||
AY033495.1 | - | 9 | 1919 | |||
BX089867.1 | - | 1 | 225 |