Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282618.1 | NP_001269547.1 | 1 | 286 | ||
NM_002070.3 | NP_002061.1 | 1 | 286 | |||
GenBank mRNA | AF493906.1 | - | 1 | 107 | ||
AK055574.1 | - | 1 | 239 | |||
AK304473.1 | - | 1 | 51 | |||
AY677118.1 | - | 1 | 107 | |||
BC012138.1 | - | 1 | 267 | 4562585 | ||
BC014627.2 | - | 1 | 224 | 2963802 | ||
CR456783.1 | - | 1 | 107 | |||
HY037331.1 | - | 1 | 286 | |||
J03004.1 | - | 1 | 184 | |||
X04828.1 | - | 1 | 230 |