Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF280B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080764.3 | NP_542942.2 | 4 | 1507 | ||
NR_130642.1 | 4 | 1507 | ||||
XM_011529893.2 | XP_011528195.1 | 5 | 1546 | |||
XM_011529895.2 | XP_011528197.1 | 3 | 1418 | |||
XM_011529896.2 | XP_011528198.1 | 3 | 1140 | |||
XM_011529897.2 | XP_011528199.1 | 3 | 1065 | |||
GenBank mRNA | BC040020.2 | - | 4 | 1053 | 5785297 | |
CR456614.1 | - | 1 | 783 |