Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080827.1 | NP_543017.1 | 3 | 466 | ||
GenBank mRNA | AF411861.1 | - | 3 | 466 |