Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080832.2 | NP_543022.1 | 2 | 1405 | ||
GenBank mRNA | AK290105.1 | - | 1 | 970 | ||
AK297327.1 | - | 3 | 1078 | |||
AL122118.1 | - | 1 | 920 | |||
BC063113.1 | - | 2 | 1183 | 6452933 |