Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080869.1 | NP_543145.1 | 3 | 275 | ||
GenBank mRNA | AY037803.1 | - | 3 | 275 | ||
AY358678.1 | - | 3 | 261 |