Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130809.4 | NP_570721.1 | 9 | 1700 | ||
XM_006714529.3 | XP_006714592.1 | 9 | 1908 | |||
GenBank mRNA | AJ427339.1 | - | 9 | 1653 | ||
AJ515428.1 | - | 9 | 1653 | |||
AJ515429.1 | - | 9 | 1653 | |||
AK057437.1 | - | 9 | 1637 | |||
AK090486.1 | - | 8 | 1525 | |||
AL831963.1 | - | 9 | 1662 | |||
AL832056.1 | - | 9 | 1663 | |||
AL832600.2 | - | 8 | 5440 | |||
BC017066.1 | - | 9 | 1637 | 3913311 |