Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031732.2 | NP_001026902.1 | 10, 11 | 1795 | ||
NM_133370.2 | NP_588611.2 | 9, 10 | 1741 | |||
XM_005265706.2 | XP_005265763.1 | 10, 11 | 1795 | |||
XM_005265707.2 | XP_005265764.1 | 10, 11 | 1929 | |||
XM_005265708.2 | XP_005265765.1 | 9, 10 | 1741 | |||
XM_017008832.1 | XP_016864321.1 | 10, 11 | 1929 | |||
XM_017008833.1 | XP_016864322.1 | 9, 10 | 1875 | |||
GenBank mRNA | AB075846.1 | - | 7, 8 | 701 | ||
AK056430.1 | - | 9, 10 | 1726 | |||
AK098515.1 | - | 7, 8 | 1081 | |||
AL832559.1 | - | 7, 8 | 1260 | |||
BC041119.1 | - | 10, 11 | 1725 | 5541053 | ||
BC053863.1 | - | 9, 10 | 1406 | 4933750 |