Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159587.2 | NP_001153059.1 | 2, 3 | 371 | ||
NM_001159588.2 | NP_001153060.1 | 2, 3 | 371 | |||
NM_133493.4 | NP_598000.2 | 2, 3 | 371 | |||
XM_005248659.3 | XP_005248716.1 | 3, 4 | 518 | |||
XM_011535473.1 | XP_011533775.1 | 2, 3 | 216 | |||
GenBank mRNA | AF410459.1 | - | 2, 3 | 352 | ||
AK095888.1 | - | 2, 3 | 305 | |||
AK313636.1 | - | 2, 3 | 351 | |||
AY149920.1 | - | 2, 3 | 240 | |||
AY788891.1 | - | 2, 3 | 265 | |||
BX641095.1 | - | 2, 3 | 258 | |||
EF553520.1 | - | 2, 3 | 270 |