Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297755.1 | NP_001284684.1 | 1 | 330 | ||
NM_001297756.1 | NP_001284685.1 | 1 | 330 | |||
NM_001297757.1 | NP_001284686.1 | 1 | 330 | |||
NM_001297758.1 | NP_001284687.1 | 1 | 330 | |||
NM_001297759.1 | NP_001284688.1 | 1 | 330 | |||
NM_133636.3 | NP_598375.2 | 1 | 330 | |||
NR_123737.1 | 1 | 330 | ||||
XM_005262711.1 | XP_005262768.1 | 1 | 302 | |||
XM_005262713.2 | XP_005262770.1 | 1 | 292 | |||
XM_006714076.2 | XP_006714139.1 | 1 | 292 | |||
XM_011531580.2 | XP_011529882.1 | 1 | 292 | |||
XM_017007679.1 | XP_016863168.1 | 1 | 302 | |||
XM_017007680.1 | XP_016863169.1 | 1 | 292 | |||
XM_017007681.1 | XP_016863170.1 | 1 | 292 | |||
XM_017007682.1 | XP_016863171.1 | 1 | 270 | |||
GenBank mRNA | AF436845.1 | - | 1 | 291 | ||
AK128665.1 | - | 1 | 301 | |||
AK308395.1 | - | 1 | 270 | |||
AK312725.1 | - | 1 | 293 | |||
BC015428.1 | - | 1 | 303 | |||
BC095473.1 | - | 1 | 238 | |||
BC111502.1 | - | 1 | 251 | |||
DB081240.1 | - | 1 | 330 | |||
KU178845.1 | - | 1 | 140 | |||
KU178846.1 | - | 1 | 140 |