Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DQX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133637.2 | NP_598376.2 | 4 | 947 | ||
XM_011532644.1 | XP_011530946.1 | 3 | 626 | |||
XM_011532645.1 | XP_011530947.1 | 1 | 59 | |||
XM_011532646.2 | XP_011530948.1 | 4 | 947 | |||
XM_017003520.1 | XP_016859009.1 | 2 | 400 | |||
GenBank mRNA | AK074337.1 | - | 4 | 854 | ||
AK092420.1 | - | 4 | 948 | |||
BC075018.2 | - | 2 | 474 | 30915408 |