Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138328.2 | NP_612201.1 | 3 | 176 | ||
XM_006721733.3 | XP_006721796.1 | 2 | 153 | |||
XM_006721734.3 | XP_006721797.1 | 2 | 545 | |||
XM_017024272.1 | XP_016879761.1 | 3 | 181 | |||
XM_017024273.1 | XP_016879762.1 | 2 | 156 | |||
XM_017024274.1 | XP_016879763.1 | 2 | 484 | |||
GenBank mRNA | AJ313480.1 | - | 3 | 176 | ||
BC100975.1 | - | 3 | 175 | 40005244 | ||
BC100976.1 | - | 3 | 176 | |||
BC100977.1 | - | 2 | 152 | 40005246 |