Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018050.3 | NP_001018060.1 | 1, 2 | 507 | ||
NM_001018052.3 | NP_001018062.1 | 1, 2 | 507 | |||
NM_001282884.1 | NP_001269813.1 | 2, 3 | 235 | |||
NM_001282885.1 | NP_001269814.1 | 2, 3 | 194 | |||
NM_138338.4 | NP_612211.1 | 2, 3 | 201 | |||
XM_011529993.1 | XP_011528295.1 | 1, 2 | 415 | |||
XM_017028641.1 | XP_016884130.1 | 1, 2 | 415 | |||
GenBank mRNA | AB051452.1 | - | 1, 2 | 362 | ||
AK026524.1 | - | 2, 3 | 181 | |||
AK129663.1 | - | 2, 3 | 184 | |||
AY092087.1 | - | 2, 3 | 179 | |||
BC017248.2 | - | 1, 2 | 394 | 4892259 | ||
BC088367.1 | - | 1, 2 | 370 | 30385483 | ||
BI754673.1 | - | 1, 2 | 390 | |||
BU173504.1 | - | 1, 2 | 149 | |||
BU928882.1 | - | 512 | ||||
BX331353.2 | - | 1, 2 | 473 | |||
BX367405.2 | - | 1, 2 | 428 | |||
CB126821.1 | - | 1, 2 | 480 | |||
CR456459.1 | - | 2, 3 | 235 | |||
DA449644.1 | - | 2, 3 | 216 | |||
DA504810.1 | - | 2, 3 | 183 |