Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138342.3 | NP_612351.2 | 5 | 672 | ||
XM_006718937.1 | XP_006719000.1 | 5 | 612 | |||
GenBank mRNA | AF173889.1 | - | 4 | 299 | ||
AK074703.1 | - | 5 | 557 | |||
AK074855.1 | - | 5 | 542 | |||
AK074947.1 | - | 5 | 496 | |||
AY358497.1 | - | 5 | 552 | |||
BC040641.1 | - | 5 | 634 | 3849776 | ||
BX640934.1 | - | 2 | 788 |