Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289034.1 | NP_001275963.1 | 3 | 782 | ||
NM_138343.3 | NP_612352.1 | 3 | 548 | |||
NM_201521.2 | NP_958929.1 | 3 | 548 | |||
NM_201522.2 | NP_958930.1 | 4 | 964 | |||
NM_201523.2 | NP_958931.1 | 3 | 838 | |||
XM_011514989.1 | XP_011513291.1 | 3 | 897 | |||
XM_017011457.1 | XP_016866946.1 | 3 | 548 | |||
GenBank mRNA | AK055293.1 | - | 4 | 926 | ||
AK056393.1 | - | 3 | 508 | |||
AK094102.1 | - | 3 | 804 | |||
AK127018.1 | - | 6 | 1813 | |||
AK127894.1 | - | 3 | 521 | |||
BC012357.1 | - | 3 | 489 | |||
BC080637.1 | - | 3 | 488 | 6573021 | ||
BC103727.1 | - | 3 | 486 | 6157949 | ||
BX640717.1 | - | 3 | 745 | |||
DA782235.1 | - | 3 | 548 |