Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145023.1 | NP_001138495.1 | 4 | 524 | ||
NM_138355.3 | NP_612364.2 | 4 | 524 | |||
XM_005257776.4 | XP_005257833.1 | 4 | 504 | |||
XM_017025286.1 | XP_016880775.1 | 4 | 452 | |||
GenBank mRNA | AK290646.1 | - | 4 | 427 | ||
AK303835.1 | - | 4 | 480 | |||
BC002980.1 | - | 4 | 467 | |||
BC010408.2 | - | 4 | 438 | 3945715 | ||
BC017317.2 | - | 4 | 437 | 3832079 |