Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVB12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304547.1 | NP_001291476.1 | 4, 5 | 512 | ||
NM_138401.3 | NP_612410.1 | 3, 4 | 585 | |||
GenBank mRNA | AK289487.1 | - | 3, 4 | 385 | ||
BC007883.2 | - | 3, 4 | 321 | 3688605 | ||
BC011840.2 | - | 3, 4 | 329 | 4053084 | ||
BG684013.1 | - | 4, 5 | 477 |