Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP140L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308162.1 | NP_001295091.1 | 10 | 920 | ||
NM_138402.4 | NP_612411.4 | 11 | 996 | |||
XM_011512190.2 | XP_011510492.1 | 11 | 1033 | |||
XM_011512191.1 | XP_011510493.1 | 11 | 1033 | |||
XM_017005294.1 | XP_016860783.1 | 11 | 1033 | |||
XM_017005295.1 | XP_016860784.1 | 11 | 1033 | |||
XM_017005296.1 | XP_016860785.1 | 11 | 982 | |||
XM_017005297.1 | XP_016860786.1 | 10 | 999 | |||
XM_017005299.1 | XP_016860788.1 | 10 | 947 | |||
XM_017005300.1 | XP_016860789.1 | 11 | 982 | |||
XM_017005301.1 | XP_016860790.1 | 11 | 1033 | |||
GenBank mRNA | AK023116.1 | - | 6 | 393 | ||
AU130836.1 | - | 6 | 393 | |||
BQ232706.1 | - | 5 | 316 | |||
CR987282.1 | - | 1 | 74 | |||
KF419365.1 | - | 10 | 830 |