Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138410.3 | NP_612419.1 | 5 | 1020 | ||
NM_181472.2 | NP_852137.1 | 4 | 921 | |||
GenBank mRNA | AF479263.2 | - | 5 | 1020 | ||
AK308878.1 | - | 4 | 619 | |||
BC010116.2 | - | 5 | 802 | 3636045 |