Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGF29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138414.2 | NP_612423.1 | 4, 5 | 406 | ||
XM_017022894.1 | XP_016878383.1 | 4, 5 | 419 | |||
GenBank mRNA | AK057008.1 | - | 4, 5 | 365 | ||
BC011981.1 | - | 4, 5 | 397 | 3863665 |