Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005059.3 | NP_005050.2 | 3 | 773 | ||
NM_134441.2 | NP_604390.1 | 2 | 672 | |||
XM_017015011.1 | XP_016870500.1 | 3 | 1041 | |||
GenBank mRNA | AF057706.1 | - | 1 | 83 | ||
BG206251.1 | - | 1 | 411 | |||
BX094853.1 | - | 2 | 501 |