Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM122A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138333.4 | NP_612206.4 | 1 | 521 | ||
GenBank mRNA | AK126379.1 | - | 1 | 462 | ||
BC013062.1 | - | 1 | 472 | 2901027 | ||
BI757526.1 | - | 1 | 478 |