Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146726.1 | NP_001140198.1 | 2 | 342 | ||
NM_138379.2 | NP_612388.2 | 2 | 342 | |||
XM_011534694.2 | XP_011532996.1 | 2 | 313 | |||
XM_017010021.1 | XP_016865510.1 | 2 | 313 | |||
GenBank mRNA | AK301337.1 | - | 2 | 313 | ||
AK312746.1 | - | 2 | 312 | |||
BC008988.1 | - | 2 | 342 | 4184237 | ||
JX049980.1 | - | 2 | 285 | |||
KU178832.1 | - | 2 | 285 | |||
KU178833.1 | - | 2 | 285 |