Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142310.1 | NP_001135782.1 | 3 | 545 | ||
NM_001142311.1 | NP_001135783.1 | 2 | 459 | |||
NM_001142312.1 | NP_001135784.1 | 2 | 453 | |||
NM_138390.3 | NP_612399.1 | 2 | 455 | |||
XM_006712851.1 | XP_006712914.1 | 2 | 659 | |||
XM_011512173.2 | XP_011510475.1 | 2 | 850 | |||
GenBank mRNA | AK091582.1 | - | 2 | 402 | ||
AK313536.1 | - | 2 | 419 | |||
BC008604.1 | - | 2 | 411 | 4180869 | ||
CR933667.1 | - | 2 | 504 | |||
DA392602.1 | - | 2 | 453 | |||
DA718294.1 | - | 3 | 492 |