Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142650.1 | NP_001136122.1 | 3 | 358 | ||
NM_138394.3 | NP_612403.2 | 2 | 614 | |||
XM_005264639.3 | XP_005264696.2 | 2 | 611 | |||
XM_005264640.3 | XP_005264697.2 | 3 | 357 | |||
XM_011533165.2 | XP_011531467.1 | 2 | 611 | |||
XM_011533166.2 | XP_011531468.1 | 2 | 611 | |||
GenBank mRNA | AK000155.1 | - | 3 | 262 | ||
AK291462.1 | - | 2 | 479 | |||
AK292259.1 | - | 3 | 351 | |||
AY236962.1 | - | 2 | 443 | |||
AY236963.2 | - | 3 | 203 | |||
BC008217.1 | - | 3 | 251 | 3449607 | ||
BC017480.1 | - | 2 | 460 | 3502860 | ||
DA760455.1 | - | 4 | 294 | |||
KU178839.1 | - | 2 | 204 | |||
KU178840.1 | - | 2 | 204 | |||
KU178841.1 | - | 2 | 204 |