Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MVB12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304547.1 | NP_001291476.1 | 7 | 820 | ||
NM_138401.3 | NP_612410.1 | 6 | 893 | |||
GenBank mRNA | AK057057.1 | - | 7 | 1378 | ||
AK289487.1 | - | 6 | 693 | |||
BC007883.2 | - | 6 | 629 | 3688605 | ||
BC011840.2 | - | 6 | 637 | 4053084 | ||
BG684013.1 | - | 7 | 785 | |||
BY996463.1 | - | 4 | 454 |