Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP140L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308162.1 | NP_001295091.1 | 10, 11 | 971 | ||
NM_138402.4 | NP_612411.4 | 11, 12 | 1047 | |||
XM_011512190.2 | XP_011510492.1 | 11, 12 | 1084 | |||
XM_017005294.1 | XP_016860783.1 | 11, 12 | 1084 | |||
XM_017005295.1 | XP_016860784.1 | 11, 12 | 1084 | |||
XM_017005296.1 | XP_016860785.1 | 11, 12 | 1033 | |||
XM_017005297.1 | XP_016860786.1 | 10, 11 | 1050 | |||
XM_017005299.1 | XP_016860788.1 | 10, 11 | 998 | |||
XM_017005300.1 | XP_016860789.1 | 11, 12 | 1033 | |||
GenBank mRNA | AK023116.1 | - | 6, 7 | 444 | ||
AU130836.1 | - | 6, 7 | 444 | |||
BQ232706.1 | - | 5, 6 | 367 | |||
KF419365.1 | - | 10, 11 | 881 |